missing translation for 'onlineSavingsMsg'
Learn More

Isophorone 97.0+%, TCI America™

Catalog No. I0151500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0151500ML 500 mL
I015125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0151500ML Supplier TCI America Supplier No. I0151500ML

Chemical Identifiers

CAS 78-59-1
Molecular Formula C9H14O
Molecular Weight (g/mol) 138.21
MDL Number MFCD00001584
InChI Key HJOVHMDZYOCNQW-UHFFFAOYSA-N
Synonym isophorone, isoacetophorone, isoforone, isoforon, izoforon, isooctopherone, 3,5,5-trimethyl-2-cyclohexen-1-one, alpha-isophorone, 2-cyclohexen-1-one, 3,5,5-trimethyl, 1,1,3-trimethyl-3-cyclohexene-5-one
PubChem CID 6544
ChEBI CHEBI:34800
IUPAC Name 3,5,5-trimethylcyclohex-2-en-1-one
SMILES CC1=CC(=O)CC(C)(C)C1

Specifications

Melting Point -8°C
Color Yellow
Boiling Point 215°C
Quantity 500 mL
Formula Weight 138.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Isophorone
EINECSNumber (3)-2381&(3)-2389
RTECSNumber GW7700000
TSCA Yes