missing translation for 'onlineSavingsMsg'
Learn More

Isopentyl butyrate, 98%

Catalog No. AAB2110322
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2110322 100 g
AAB2110336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2110322 Supplier Thermo Scientific Chemicals Supplier No. B2110322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 106-27-4 | C9H18O2 | 158.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106-27-4
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD00044888
InChI Key PQLMXFQTAMDXIZ-UHFFFAOYSA-N
Synonym isoamyl butyrate, isopentyl butyrate, isopentyl butanoate, isoamyl butanoate, butanoic acid, 3-methylbutyl ester, 3-methylbutyl butyrate, butyric acid, isopentyl ester, isoamyl-n-butyrate, isopentyl alcohol, butyrate, isoamyl butylate
PubChem CID 7795
ChEBI CHEBI:87422
IUPAC Name 3-methylbutyl butanoate
SMILES CCCC(=O)OCCC(C)C

Specifications

Melting Point -73°C
Density 0.88
Boiling Point 178°C
Flash Point 59°C (138°F)
Odor Fruit-like
Refractive Index 1.411
Quantity 100 g
UN Number UN2620
Merck Index 14,5115
Formula Weight 158.24
Percent Purity 98%
Chemical Name or Material Isopentyl butyrate
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMYL BUTYRATES
EINECSNumber 203-380-8
RTECSNumber ET5034000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only