missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopentyl acetate, ≥99.7% (GC), MilliporeSigma™ Supelco™
Isopentyl acetate is an alarm pheromone present in honey bees, which may affect the brain and hence, provoke a quick defensive response, thereby influencing their behavior for a longer period of time.
Supplier: Merck Emd Millipore 798575ML
Description
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal CareSpecifications
| 680°F | |
| 5 mmHg (25°C) | |
| 0.876 g/mL (at 25°C) | |
| Neat | |
| MFCD00008946 | |
| 142°C (756 mmHg) | |
| Analytical Standard | |
| 130.18 | |
| Analytical Standard |
| 123-92-2 | |
| 4.5 (vs. air) | |
| 33°C - Closed Cup | |
| CH3 COOCH2 CH2 CH(CH3)2 | |
| -78°C | |
| Acetic acid 3-methylbutyl ester; Isoamyl acetate | |
| 5 mL | |
| ≥99.7% (GC) |