missing translation for 'onlineSavingsMsg'
Learn More

Isopentyl acetate, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7188022
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020619 1 mL
111019345 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020619 Supplier Merck Emd Millipore Supplier No. 798571ML
Only null left

Isopentyl acetate is an alarm pheromone present in honey bees, which may affect the brain and hence, provoke a quick defensive response, thereby influencing their behavior for a longer period of time.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Ignition Point 680°F
CAS 123-92-2
Vapor Pressure 5 mmHg (25°C)
Vapor Density 4.5 (vs. air)
Density 0.876 g/mL (at 25°C)
Flash Point 33°C - Closed Cup
Physical Form Neat
Linear Formula CH3 COOCH2 CH2 CH(CH3)2
MDL Number MFCD00008946
Melting Point -78°C
Boiling Point 142°C (756 mmHg)
Synonym Acetic acid 3-methylbutyl ester; Isoamyl acetate
Type Analytical Standard
Quantity 1 mL
Formula Weight 130.18
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less