missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Isonipecotic Acid, 99%

Numéro de catalogue. p-3737953
Modifier l'affichage
Click to view available options
Quantité:
25g
100g
500g
Emballage:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC122620250 25g Glass bottle
AC122621000 100g Glass bottle
AC122625000 500g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AC122620250 Fournisseur Thermo Scientific™ Code fournisseur 122620250

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 498-94-2
Formule moléculaire C6H11NO2
Poids moléculaire (g/mol) 129.16
Numéro MDL MFCD00006004
Clé InChI SRJOCJYGOFTFLH-UHFFFAOYSA-N
Synonyme isonipecotic acid, 4-piperidinecarboxylic acid, 4-carboxypiperidine, hexahydroisonicotinic acid, dl-isopipecolinic acid, isonicotinic acid, hexahydro, h-inp-oh, acide isonipecotique, 4-hexahydroisonicotinic acid, unii-m5tzp1rwie
PubChem CID 3773
Nom de l’IUPAC piperidine-4-carboxylic acid
SOURIRES OC(=O)C1CCNCC1

Spécifications

Nom ou substance chimique Isonipecotic acid
Perte à la dessication 1% max. (120°C, 4 hrs)
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 99%
Emballage Glass bottle
Quantité 25g
Beilstein 22, V,1, 244
Indice de Merck 15, 5235
Renseignements sur la solubilité (10% in water) Clear to slightly hazy
Poids de formule 129.16
Pourcentage de pureté 99%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-872-3

RUO – Research Use Only