missing translation for 'onlineSavingsMsg'
Learn More

Isonicotinoyl chloride hydrochloride, 97%

Catalog No. AAA1086309
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1086309 10 g
AAA1086318 50 g
AAA1086330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1086309 Supplier Thermo Scientific Chemicals Supplier No. A1086309

CAS: 39178-35-3 | C6H5Cl2NO | 178.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39178-35-3
Molecular Formula C6H5Cl2NO
Molecular Weight (g/mol) 178.01
MDL Number MFCD00012830
InChI Key BNTRVUUJBGBGLZ-UHFFFAOYSA-N
Synonym isonicotinoyl chloride hydrochloride, pyridine-4-carbonyl chloride hydrochloride, isonicotinoylchloride hydrochloride, 4-pyridinecarbonyl chloride hydrochloride, 4-pyridinecarbonyl chloride, hydrochloride, isonicotinoylchloridehcl, pubchem23974, acmc-1bn6p, ksc223m5d, 4-pyridinecarbonyl chloride-hcl
PubChem CID 12262826
IUPAC Name pyridine-4-carbonyl chloride;hydrochloride
SMILES [H+].[Cl-].ClC(=O)C1=CC=NC=C1

Specifications

Melting Point 154°C to 162°C
Quantity 10 g
UN Number UN3261
Beilstein 3626052
Sensitivity Hygroscopic
Formula Weight 178.01
Percent Purity 98%
Chemical Name or Material Isonicotinoyl chloride hydrochloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 254-331-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only