missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Isomaltotriose, 97%

Catalog No. AAJ64186ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64186ME 500 mg
AAJ64186MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64186ME Supplier Thermo Scientific Chemicals Supplier No. J64186ME

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3371-50-4 | C18H32O16 | 504.438 g/mol

Isomaltotriose is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isomaltotriose is used as pharmaceutical intermediate.

Solubility
Soluble in water

Notes
Air sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 3371-50-4
Molecular Formula C18H32O16
Molecular Weight (g/mol) 504.438
MDL Number MFCD00070623
InChI Key FBJQEBRMDXPWNX-IGSOQDMUSA-N
Synonym isomaltotriose, 3r,4s,5s,6r-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-2-yl oxy methyl oxane-2,3,4,5-tetrol, isomaltotriose, dp3, glc a1-6 glc a1-6 glc, isomaltotriose tlc, isomaltotriose, dp3, analytical standard, alpha-d-glucosyl-1->6-alpha-d-glucosyl-1->6-d-glucose, 6-o-6-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-d-glucopyranose, d-glucose, o-.alpha.-d-glucopyranosyl-1->6-o-.alpha.-d-glucopyranosyl-1->6, 3r,4s,5s,6r-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxymethyl oxane-2,3,4,5-tetrol
PubChem CID 133126476
IUPAC Name (3S,4R,5R,6S)-6-[[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-[[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxan-2-yl]oxymethyl]oxane-2,3,4,5-tetrol
SMILES C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OCC3C(C(C(C(O3)O)O)O)O)O)O)O)O)O)O)O

Specifications

Color White
Quantity 500 mg
Solubility Information Soluble in water
Formula Weight 504.44
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Isomaltotriose
EINECSNumber 222-149-2
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only