missing translation for 'onlineSavingsMsg'
Learn More

Isochroman 98.0+%, TCI America™

Catalog No. I051425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I051425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I051425G Supplier TCI America Supplier No. I051425G

Chemical Identifiers

CAS 493-05-0
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00006913
InChI Key HEBMCVBCEDMUOF-UHFFFAOYSA-N
Synonym isochroman, isochromane, 3,4-dihydro-1h-2-benzopyran, 1h-2-benzopyran, 3,4-dihydro, 3,4-dihydro-1h-benzo c pyran, pubchem20553, acmc-209kfu, isochroman 10g, ksc237m1l
PubChem CID 96266
ChEBI CHEBI:33225
IUPAC Name 3,4-dihydro-1H-2-benzopyran
SMILES C1CC2=CC=CC=C2CO1

Specifications

Color Yellow
Boiling Point 110°C
Quantity 25 g
Formula Weight 134.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isochroman