missing translation for 'onlineSavingsMsg'
Learn More

Isobutyric Anhydride 95.0+%, TCI America™

Catalog No. I011125ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I011125ML 25 mL
I0111500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I011125ML Supplier TCI America Supplier No. I011125ML
Only null left

Chemical Identifiers

CAS 97-72-3
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD00008913
InChI Key LSACYLWPPQLVSM-UHFFFAOYSA-N
Synonym isobutyric anhydride, isobutyric acid anhydride, propanoic acid, 2-methyl-, anhydride, 2-methylpropanoic anhydride, isobutyricanhydride, isobutyryl anhydride, 2-methylpropionic anhydride, unii-n85a80fjdt, n85a80fjdt, 2-methylpropanoic acid anhydride
PubChem CID 7346
ChEBI CHEBI:84261
IUPAC Name 2-methylpropanoyl 2-methylpropanoate
SMILES CC(C)C(=O)OC(=O)C(C)C

Specifications

Color Yellow
Boiling Point 182°C
Quantity 25 mL
UN Number 3265
Formula Weight 158.2
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Isobutyric Anhydride
EINECSNumber (2)-4182
TSCA Yes