missing translation for 'onlineSavingsMsg'
Learn More

Isobutyramide, 99%

Catalog No. AAB2164114
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2164114 25 g
AAB2164122 100 g
AAB2164136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2164114 Supplier Thermo Scientific Chemicals Supplier No. B2164114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 563-83-7 | C4H9NO | 87.122 g/mol

Isobutyramide plays an important role in chemical grafting of human serum albumin during the synthesis of sequentially assembled protein capsules. It is used in pharmaceutical manufacturing and in cellular differentiation. It finds application in the transcription of human gamma-globin gene and murine embryonic epsilon(y)-globin gene. Further, it is in the treatment of beta-thalassemia and sickle cell disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Incompatible with acids, bases, strong oxidizing agents and strong reducing agents.

Chemical Identifiers

CAS 563-83-7
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.122
MDL Number MFCD00008019
InChI Key WFKAJVHLWXSISD-UHFFFAOYSA-N
Synonym isobutyramide, propanamide, 2-methyl, 2-methylpropionamide, isobutyrimidic acid, isobutylamide, isopropylformamide, dimethylacetoamide, unii-82uoe7b38z, dimethylacetamid, c-isopropylformamide
PubChem CID 68424
IUPAC Name 2-methylpropanamide
SMILES CC(C)C(=O)N

Specifications

Melting Point 127°C to 129°C
Density 1.013
Boiling Point 216°C to 220°C
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight 87.12
Percent Purity 99%
Chemical Name or Material Isobutyramide
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 209-265-9
RTECSNumber TX1475000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only