missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutyl Mercaptan 98.0+%, TCI America™
Spécifications
Spécifications
| Color | Yellow |
| Boiling Point | 88°C |
| Quantity | 500 mL |
| UN Number | 2347 |
| Formula Weight | 90.18 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Isobutyl Mercaptan |
Sécurité et manipulation
| EINECSNumber | (2)-0464 |
| RTECSNumber | TZ7630000 |
| TSCA | Yes |