missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutyl Isothiocyanate 97.0+%, TCI America™
Supplier: TCI America I018625ML
Specifications
| Isobutyl Isothiocyanate | |
| Yellow | |
| C5H9NS | |
| 25 mL | |
| isobutyl isothiocyanate, i-butyl isothiocyanate, isobutylisothiocyanate, 2-methylpropyl isothiocyanate, isothiocyanic acid, isobutyl ester, 1-isothiocyanato-2-methyl-propane, unii-a498zc6z4f, isothiocyanic acid isobutyl ester, propane, 1-isothiocyanato-2-methyl, propane, 1-isothiocyanato-2-methyl-9ci | |
| CC(C)CN=C=S | |
| 115.194 | |
| 115.19 | |
| Liquid |
| 591-82-2 | |
| 164°C | |
| MFCD00022062 | |
| 1992 | |
| NSDDRJXKROCWRZ-UHFFFAOYSA-N | |
| 1-isothiocyanato-2-methylpropane | |
| 68960 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 591-82-2 | |
| 115.194 | |
| NSDDRJXKROCWRZ-UHFFFAOYSA-N | |
| 68960 | |
| CC(C)CN=C=S |
| C5H9NS | |
| MFCD00022062 | |
| isobutyl isothiocyanate, i-butyl isothiocyanate, isobutylisothiocyanate, 2-methylpropyl isothiocyanate, isothiocyanic acid, isobutyl ester, 1-isothiocyanato-2-methyl-propane, unii-a498zc6z4f, isothiocyanic acid isobutyl ester, propane, 1-isothiocyanato-2-methyl, propane, 1-isothiocyanato-2-methyl-9ci | |
| 1-isothiocyanato-2-methylpropane |
Safety and Handling
RTECSNumber : NX8780100
TSCA : No