missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Isothiocyanate 97.0+%, TCI America™

Catalog No. p-7131260
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I01865ML 5 mL
I018625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I01865ML Supplier TCI America Supplier No. I01865ML
Only null left

Chemical Identifiers

CAS 591-82-2
Molecular Formula C5H9NS
Molecular Weight (g/mol) 115.194
MDL Number MFCD00022062
InChI Key NSDDRJXKROCWRZ-UHFFFAOYSA-N
Synonym isobutyl isothiocyanate, i-butyl isothiocyanate, isobutylisothiocyanate, 2-methylpropyl isothiocyanate, isothiocyanic acid, isobutyl ester, 1-isothiocyanato-2-methyl-propane, unii-a498zc6z4f, isothiocyanic acid isobutyl ester, propane, 1-isothiocyanato-2-methyl, propane, 1-isothiocyanato-2-methyl-9ci
PubChem CID 68960
IUPAC Name 1-isothiocyanato-2-methylpropane
SMILES CC(C)CN=C=S

Specifications

Color Yellow
Boiling Point 164°C
Quantity 5 mL
UN Number 1992
Formula Weight 115.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Isothiocyanate
RTECSNumber NX8780100
TSCA No