missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl acrylate, 99%, stab with 100ppm 4-methoxyphenol

Catalog No. AAL12739AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL12739AE 100 mL
AAL12739AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL12739AE Supplier Thermo Scientific Chemicals Supplier No. L12739AE

CAS: 106-63-8 | C7H12O2 | 128.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106-63-8
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00042865
InChI Key CFVWNXQPGQOHRJ-UHFFFAOYSA-N
Synonym isobutyl acrylate, isobutyl propenoate, isobutyl 2-propenoate, 2-propenoic acid, 2-methylpropyl ester, acrylic acid, isobutyl ester, 2-methylpropyl acrylate, isobutylester kyseliny akrylove, propenoic acid, isobutyl ester, ccris 4828, hsdb 610
PubChem CID 7819
IUPAC Name 2-methylpropyl prop-2-enoate
SMILES CC(C)COC(=O)C=C

Specifications

Melting Point -61°C
Density 0.888
Boiling Point 139°C
Flash Point 32°C (89°F)
Refractive Index 1.415
Linear Formula CH2=CHCO2CH2CH(CH3)2
Quantity 100 mL
UN Number UN2527
Beilstein 1749388
Formula Weight 128.17
Percent Purity 99%
Chemical Name or Material Isobutyl acrylate, stabilized with 100ppm 4-methoxyphenol
Show More Show Less
Hazard Category H226-H312+H332-H315-H317-H319
Hazard Statement GHS H Statement
H226-H312-H332-H315-H317
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ISOBUTYL ACRYLATE, STABILIZED
EINECSNumber 203-417-8
RTECSNumber AT2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only