missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl Éther (stabilisé avec BHT) 98,0+%, TCI America™

Numéro de catalogue. I0086500ML
Modifier l'affichage
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
I0086500ML 500 mL
I008625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. I0086500ML Fournisseur TCI America Code fournisseur I0086500ML

Identifiants chimiques

CAS 544-01-4
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.285
MDL Number MFCD00008947
InChI Key AQZGPSLYZOOYQP-UHFFFAOYSA-N
Synonym diisopentyl ether, isoamyl ether, isopentyl ether, diisoamyl ether, isoamyl oxide, butane, 1,1'-oxybis 3-methyl, di-3-methylbutyl ether, di-iso-amyl ether, 1,1'-oxybis 3-methylbutane, 1,1'-oxylbis 3-methylbutane
PubChem CID 10989
IUPAC Name 3-méthyl-1-(3-méthylbutoxy)butane
SMILES CC(C)CCOCCC(C)C

Spécifications

Color Incolore
Boiling Point 173°C
Quantity 500 mL
UN Number 3271
Formula Weight 158.29
Percent Purity ≥98.0% (GC)
Physical Form Liquide
Chemical Name or Material Isoamyl Ether (stabilized with BHT)
EINECSNumber (2)-0365
RTECSNumber EK5433750
TSCA Yes