missing translation for 'onlineSavingsMsg'
Learn More
Learn More
iso-Butyl Alcohol, ACS, 99%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor B1186500MLGL
Specifications
| Iso-Butyl Alcohol | |
| 1 | |
| 500 mL | |
| 1e-05 | |
| 0.001 | |
| 0.99 |
| 78-83-1 | |
| 108°C | |
| Amber Glass Bottle | |
| 0.0005 meq/g | |
| 74.12 | |
| Reagent |
Chemical Identifiers
| 78-83-1 | |
| 74.12 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.