missing translation for 'onlineSavingsMsg'
Learn More

Isethionic acid sodium salt, 98%

Catalog No. AAA120540I
Change view
Click to view available options
Quantity:
1000 g
5000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA120540I 5000 g
AAA120540B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA120540I Supplier Thermo Scientific Chemicals Supplier No. A120540I

CAS: 1562-00-1 | C2H5NaO4S | 148.108 g/mol

Isethionic acid sodium salt is an amphoteric detergent used in detergent bar soaps. It makes a dense lather in addition to the lather made by the soap. It is mild on the skin, and non-drying. It works equally well in soft or hard water. It is also an anti-static agent in shampoos.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isethionic acid sodium salt is an amphoteric detergent used in detergent bar soaps. It makes a dense lather in addition to the lather made by the soap. It is mild on the skin, and non-drying. It works equally well in soft or hard water. It is also an anti-static agent in shampoos.

Solubility
Soluble in water.

Notes
Store in cool place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 1562-00-1
Molecular Formula C2H5NaO4S
Molecular Weight (g/mol) 148.108
MDL Number MFCD00007534
InChI Key LADXKQRVAFSPTR-UHFFFAOYSA-M
Synonym sodium 2-hydroxyethanesulfonate, sodium isethionate, isethionic acid sodium salt, 2-hydroxyethanesulfonic acid sodium salt, sodium hydroxyethylsulfonate, ethanesulfonic acid, 2-hydroxy-, monosodium salt, sodium 2-hydroxyethylsulfonate, sodium 2-hydroxyethanesulphonate, sodium beta-hydroxyethanesulfonate, sodium 1-hydroxy-2-ethanesulfonate
PubChem CID 517063
IUPAC Name sodium;2-hydroxyethanesulfonate
SMILES C(CS(=O)(=O)[O-])O.[Na+]

Specifications

Melting Point 191°C to 194°C
Quantity 5000 g
Beilstein 3633992
Solubility Information Soluble in water.
Formula Weight 148.11
Percent Purity 98%
Chemical Name or Material Isethionic acid sodium salt
EINECSNumber 216-343-6
RTECSNumber KI7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only