missing translation for 'onlineSavingsMsg'
Learn More

Iron Standard Stock, USP <241>, Ricca Chemical

Catalog No. R4199000500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R4199000500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R4199000500 Supplier Ricca Chemical Company Supplier No. R4199000500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7783-83-7 , 7664-93-9 , 7732-18-5
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) 482.176
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
PubChem CID 197097
IUPAC Name azanium;iron(3+);disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]

Specifications

Packaging Natural Poly Bottle
Physical Form Liquid
Color Yellow
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Analytical Standard
Quantity 500 mL
Grade Laboratory
Chemical Name or Material Iron Standard Stock USP 241
Show More Show Less