Learn More
Iron Standard, Certified, 200ppm ±2ppm (1mL = 0.2mg Fe), LabChem™
Supplier: LabChem LC157201
Specifications
| Iron Standard | |
| Poly Bottle | |
| 96.36,3.5,0.13,0.01 | |
| 1g/mL | |
| FeH20N2O14S2 | |
| Fe(NH4)2(SO4)2·6H2O | |
| UN3264 | |
| Soluble in water | |
| [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
| 392.13 | |
| 392.14 |
| 200ppm (1mL = 0.2mg Fe) | |
| 7783-85-9 , 7664-93-9 , 7722-64-7 , 7732-18-5 | |
| Passes Test | |
| Liquid | |
| 200 ppm ±2 ppm (1mL = 0.2mg Fe) | |
| MFCD00150530 | |
| Pink | |
| MQLVWQSVRZVNIP-UHFFFAOYSA-L | |
| λ²-iron(2+) diammonium hexahydrate disulfate | |
| 500 mL | |
| Certified |
Chemical Identifiers
| 7783-85-9 | |
| 392.13 | |
| MQLVWQSVRZVNIP-UHFFFAOYSA-L | |
| [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
| FeH20N2O14S2 | |
| MFCD00150530 | |
| λ²-iron(2+) diammonium hexahydrate disulfate |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature