missing translation for 'onlineSavingsMsg'
Learn More

Iron Standard, Certified, 200ppm ±2ppm (1mL = 0.2mg Fe), LabChem™

Catalog No. LC157207
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC157207 125 mL Poly Bottle
LC157201 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC157207 Supplier LabChem Supplier No. LC157207
Only null left

Chemical Identifiers

CAS 7783-85-9 , 7664-93-9 , 7722-64-7 , 7732-18-5
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Packaging Poly Bottle
CAS Max % 96.36, 3.5, 0.13, 0.01
Identification Passes Test
Density 1g/mL
Physical Form Liquid
Concentration 200 ppm ±2 ppm (1mL = 0.2mg Fe)
Linear Formula Fe(NH4)2(SO4)2·6H2O
UN Number UN3264
Color Pink
Solubility Soluble in water
Quantity 125 mL
Formula Weight 392.14
Grade Certified
Chemical Name or Material Iron Standard, 200ppm (1mL = 0.2mg Fe)
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature