missing translation for 'onlineSavingsMsg'
Learn More

Iron slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog No. AA4238222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4238222 100 g
AA4238214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4238222 Supplier Thermo Scientific Chemicals Supplier No. 04238222

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7439-89-6 | Molecular Formula: Fe | Molecular Weight: 55.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]

Specifications

Quantity 100 g
Weight ≈3.17g/slug
Form Slug
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Iron slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length
Hazard Category H500
EINECSNumber 231-096-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only