Learn More
Iron nickel oxide, tech.
Description
Iron nickel oxide is used as an adsorbent to remove hexavalent chromium ions present in the waste water by co-sedimentation method. It is also employed as an additive in membrane (i.e. polyvinylchloride/styrene-butadiene-rubber blend heterogeneous cation exchange membrane) fabrication, thereby increasing the permeability for bivalent ions compared to the monovalent type.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Brown |
| Physical Form | Powder |
| Quantity | 50 g |
| Formula Weight | 234.39 |
| Grade | Technical |
| Odor | Odorless |
| Appearance | Dark-brown powder |
| Density | 5.368 g/cm3 at 20°C |
| Chemical Name or Material | Iron nickel oxide |
Safety and Handling
| Hazard Category | H317-H350 |
| Precautionary Statement | P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c |
| EINECSNumber | 235-335-3 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only