missing translation for 'onlineSavingsMsg'
Learn More

Iron nickel oxide, tech.

Catalog No. AA8853009
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8853009 10 g
AA8853018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA8853009 Supplier Thermo Scientific Chemicals Supplier No. 08853009
Only null left

CAS: 12168-54-6 | Fe2NiO4 | 234.38 g/mol

Iron nickel oxide is used as an adsorbent to remove hexavalent chromium ions present in the waste water by co-sedimentation method. It is also employed as an additive in membrane (i.e. polyvinylchloride/styrene-butadiene-rubber blend heterogeneous cation exchange membrane) fabrication, thereby increasing the permeability for bivalent ions compared to the monovalent type.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12168-54-6
Molecular Formula Fe2NiO4
Molecular Weight (g/mol) 234.38
MDL Number MFCD00016254
InChI Key NQNBVCBUOCNRFZ-UHFFFAOYSA-N
IUPAC Name oxo[(oxoferrio)oxy]iron; oxonickel
SMILES O=[Ni].O=[Fe]O[Fe]=O

Specifications

Color Brown
Physical Form Powder
Quantity 10 g
Formula Weight 234.39
Grade Technical
Odor Odorless
Appearance Dark-brown powder
Density 5.368
Chemical Name or Material Iron nickel oxide
Hazard Category H317-H350
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
EINECSNumber 235-335-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only