missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) perchlorate hydrate, Reagent Grade

Catalog No. AA1164736
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1164736 500 g
AA1164722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1164736 Supplier Thermo Scientific Chemicals Supplier No. 01164736

CAS: 15201-61-3 | Cl3FeO12 | 354.18 g/mol

Iron(III) perchlorate hydrate is used as a starting material for the synthesis of a tris-salicylate Fe(III) complex having bidentate chelating units that allow two coordination modes. It is also used as a catalyst for synthesis of fullerene-fused lactones, α-Carbonyl furans via one-pot cyclization, fullerodioxolanes via heterocyclization, dialylated indoles via double alkylation, aryl esters via oxidative esterification and as a chemical actinometer based on photolysis of ferrioxalate in presence of polyoxometalate in aqueous solution, as an oxidant for conducting polymer nanoparticles synthesized in ionic liquid by chemical polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15201-61-3
Molecular Formula Cl3FeO12
Molecular Weight (g/mol) 354.18
MDL Number MFCD00011000
InChI Key LHOWRPZTCLUDOI-UHFFFAOYSA-K
Synonym iron iii perchlorate hydrate, perchloric acid iron iii salt hydrate, iron 3+ hydrate triperchlorate, iron iii perchlorate hydrate purple, iron iii perchlorate hydrate yellow, iron iii perchloratehydrate,reagentgrade, iron iii perchlorate hydrate, crystalline, iron iii perchlorate hydrate, crystalline, low chloride
PubChem CID 9799594
IUPAC Name iron(3+);triperchlorate;hydrate
SMILES [Fe+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Color White
Physical Form Crystalline
Quantity 500 g
Solubility Information Soluble in water.
Formula Weight 354.19 (Anhydrous)
Grade Reagent
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Iron(III) perchlorate hydrate
Hazard Category H272-H315-H319-H335
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: II
EINECSNumber 236-908-0
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only