missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) 2-ethylhexanoate diisopropoxide, 10% w/v in isopropanol, Thermo Scientific™

Catalog No. AA40583AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40583AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA40583AE Supplier Thermo Scientific Chemicals Supplier No. 40583AE

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 331720-17-3
Molecular Formula C14H32FeO4
Molecular Weight (g/mol) 320.251
MDL Number MFCD00798540
InChI Key GCDBQBNMPOKFEM-UHFFFAOYSA-N
Synonym iron iii 2-ethylhexano-isopropoxide,in isopropanol, iron 3+ 2-ethylhexanoate 2-propanolate 1:1:2, iron iii ethylhexano-isopropoxide w/v in isopropanol
PubChem CID 121233720
IUPAC Name 2-ethylhexanoic acid;iron;propan-2-ol
SMILES CCCCC(CC)C(=O)O.CC(C)O.CC(C)O.[Fe]

Specifications

Quantity 100 mL
UN Number UN1219
Sensitivity Moisture sensitive
Formula Weight 317.23
Concentration or Composition (by Analyte or Components) 10% w/v in isopropanol
Physical Form Liquid
Chemical Name or Material Iron(III) 2-ethylhexanoate diisopropoxide
Hazard Category H225-H315-H318-H335-H336
Hazard Statement GHS H Statement
H225-H361-H315-H319-H336
Highly flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
TSCA No
Recommended Storage Ambient temperatures