missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) gluconate hydrate , MilliporeSigma™ Supelco™

Catalog No. 111015277
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015277 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015277 Supplier Merck Emd Millipore Supplier No. 44948250G

Chemical Identifiers

MDL Number MFCD04037236
Synonym Ferrous gluconate hydrate

Specifications

CAS 699014-53-4
Linear Formula (C6H11O7)2 Fe · aq
Quantity 250 g
Formula Weight 446.14 (anhydrous basis)