missing translation for 'onlineSavingsMsg'
Learn More

Iron ICP Standard, 1000 ppm Fe in 3% HNO3, Ricca Chemical

$329.30 - $631.77

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
PFE1KN-100
SDS View Documents
Ricca Chemical Company
PFE1KN100
100 mL Natural LDPE Bottle
Each for $329.30
Only null left
 
PFE1KN-500
View Documents
Ricca Chemical Company
PFE1KN500
500 mL Natural Poly Bottle
Each for $631.77
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7439-89-6
55.85
XEEYBQQBJWHFJM-UHFFFAOYSA-N
CHEBI:82664
[Fe]
Fe
MFCD00010999
23925
iron

Specifications

Natural LDPE Bottle
1,000ppm Fe
97.0,5.09,0.1
Liquid
MFCD00010999
< 2
Standard
[Fe]
55.85
23925
Iron ICP Standard
7439-89-6,7697-37-2,7732-18-5
93.0,4.89,0.09
1 mL = 1 mg Fe
Fe
Colorless to Yellow
Approximately 0°C
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
100 mL
CHEBI:82664
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'