missing translation for 'onlineSavingsMsg'
Learn More

Iron ICP Standard, 1000 ppm Fe in 3% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company PFE1KN500

 View more versions of this product

Catalog No. PFE1KN-500

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Iron ICP Standard
7439-89-6,7697-37-2,7732-18-5
93.0,4.89,0.09
1 mL = 1 mg Fe
Fe
Colorless to Yellow
Approximately 0°C
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
500 mL
CHEBI:82664
Natural Poly Bottle
1,000ppm Fe
97.0,5.09,0.1
Liquid
MFCD00010999
< 2
Standard
[Fe]
55.85
23925

Chemical Identifiers

7439-89-6
55.85
XEEYBQQBJWHFJM-UHFFFAOYSA-N
CHEBI:82664
[Fe]
Fe
MFCD00010999
23925
iron
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions