missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron ICP Standard, 1000 ppm Fe in 3% HNO3, Ricca Chemical
Supplier: Ricca Chemical Company PFE1KN100
Specifications
| Iron ICP Standard | |
| 7439-89-6,7697-37-2,7732-18-5 | |
| 93.0,4.89,0.09 | |
| 1 mL = 1 mg Fe | |
| Fe | |
| Colorless to Yellow | |
| Approximately 0°C | |
| XEEYBQQBJWHFJM-UHFFFAOYSA-N | |
| iron | |
| 100 mL | |
| CHEBI:82664 |
| Natural LDPE Bottle | |
| 1,000ppm Fe | |
| 97.0,5.09,0.1 | |
| Liquid | |
| MFCD00010999 | |
| < 2 | |
| Standard | |
| [Fe] | |
| 55.85 | |
| 23925 |
Chemical Identifiers
| 7439-89-6 | |
| 55.85 | |
| XEEYBQQBJWHFJM-UHFFFAOYSA-N | |
| CHEBI:82664 | |
| [Fe] |
| Fe | |
| MFCD00010999 | |
| 23925 | |
| iron |