missing translation for 'onlineSavingsMsg'
Learn More

Iron ICP-MS Standard, 1000 ppm Fe in 3% HNO3, Ricca Chemical

Catalog No. MSFE1KN100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSFE1KN100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSFE1KN100 Supplier Ricca Chemical Company Supplier No. MSFE1KN100

Chemical Identifiers

CAS 7439-89-6 , 7697-37-2 , 7732-18-5
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Fe
CAS Min % 93.0, 4.89, 0.09
CAS Max % 97.0, 5.09, 0.1
Equivalence 1 mL = 1 mg Fe
Physical Form Liquid
Color Colorless to Yellow
pH < 2
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type ICP Single-element Standard Solution
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Iron ICP-MS Standard
Show More Show Less