missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron AA Standard, 1000 ppm Fe in 3% HNO3, Ricca Chemical
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| Concentration or Composition Notes | 1,000ppm Fe |
| CAS Min % | 93.0, 4.2, 0.68 |
| CAS Max % | 97.0, 4.37, 0.71 |
| Equivalence | 1 mL = 1 mg Fe |
| Physical Form | Liquid |
| Color | Colorless to Yellow |
| pH | < 2 |
| Melting Point | Approximately 0°C |
| Quantity | 500 mL |
| Show More |