missing translation for 'onlineSavingsMsg'
Learn More

Iron Standard, 10 ppm Fe in 3% HNO3, Ricca Chemical

Catalog No. R4183500500
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R4183500500 500 mL Natural Poly Bottle
R4183500100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R4183500500 Supplier Ricca Chemical Company Supplier No. R4183500500N

Chemical Identifiers

CAS 7439-89-6 , 7697-37-2 , 7732-18-5
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]

Specifications

Packaging Natural Poly Bottle
Odor Odorless
Assay Percent Range 10.00 ± 0.05ppm
Physical Form Liquid
Color Colorless to Yellow
Melting Point Approximately 0°C
Solubility Infinitely soluble in water
Solvent or Matrix Water
Quantity 500 mL
Chemical Name or Material Iron AA, ICP and ICP-MS Standards, 10ppm Fe in 3 to 5% Nitric Acid