missing translation for 'onlineSavingsMsg'
Learn More

Irinotecan hydrochloride trihydrate

Catalog No. AC461542500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461542500 250 mg
AC461540010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461542500 Supplier Thermo Scientific Chemicals Supplier No. 461542500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 136572-09-3 | HCl·3H2O | 677.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 136572-09-3
Molecular Formula HCl·3H2O
Molecular Weight (g/mol) 677.18
InChI Key KLEAIHJJLUAXIQ-JDRGBKBRSA-N
Synonym irinotecan hydrochloride trihydrate, irinotecan hcl trihydrate, cpt 11, unii-042laq1iis, campto, irinotecan hcl, 042laq1iis, onivyde, cpt-11
PubChem CID 60837
SMILES CCC1=C2C=C(C=CC2=NC3=C1CN4C3=CC5=C(C4=O)COC(=O)C5(CC)O)OC(=O)N6CCC(CC6)N7CCCCC7.O.O.O.Cl

Specifications

Infrared Spectrum Authentic
Quantity 250 mg
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 677.18
Percent Purity ≥96.0% (HPLC)
Chemical Name or Material Irinotecan hydrochloride trihydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling.

RUO – Research Use Only