missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iridium Standard For ICP, 1000μg/mL in 10% HCl, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$854.00 - $854.00
Chemical Identifiers
| CAS | 7439-88-5 |
|---|---|
| Molecular Formula | Ir |
| Molecular Weight (g/mol) | 192.22 |
| MDL Number | MFCD00011062 |
| InChI Key | GKOZUEZYRPOHIO-UHFFFAOYSA-N |
| PubChem CID | 23924 |
| ChEBI | CHEBI:49666 |
| IUPAC Name | iridium |
| SMILES | [Ir] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Ir | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
| 7439-88-5 | |
| 192.22 | |
| GKOZUEZYRPOHIO-UHFFFAOYSA-N | |
| CHEBI:49666 | |
| [Ir] |
| Ir | |
| MFCD00011062 | |
| 23924 | |
| iridium |
Specifications
| Triple-leached LDPE bottle | |
| Assurance Certificate of Analysis reports up to 39 elements at ppb levels. | |
| 0.1,10.0,89.9 | |
| Ir | |
| AA, ICP | |
| Certified Reference Material | |
| [Ir] | |
| iridium | |
| 125 mL | |
| CHEBI:49666 | |
| Matrix Concentration Cat. No. 10% HCl 1000μg/mL PLIR3-2Y, -2X |
| 7439-88-5 , 7647-01-0 , 7732-18-5 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 0.1,10.0,89.9 | |
| MFCD00011062 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| GKOZUEZYRPOHIO-UHFFFAOYSA-N | |
| 10% HCl | |
| 192.22 | |
| 23924 | |
| Plasma | |
| Iridium Standard |
Safety and Handling
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only