missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ IRAK1/4 Inhibitor I
Description
IRAK1/4 Inhibitor I is an IRAK4 and IRAK1 inhibitor (IC50 values are 0.2 and 0.3 μM, respectively).
Specifications
Specifications
| Chemical Name or Material | N-[1-[2-(4-Morpholinyl)ethyl]-1H-benzimidazol-2-yl]-3-nitrobenzamide |
| CAS | 509093-47-4 |
| Quantity | 50 mg |
| Target | Other Kinase Inhibitors |
| Molecular Formula | C20H21N5O4 |
| Formula Weight | 395.41 |
| Purity | >98% |