missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ IOX 1

Catalog No. 446450
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
446450 50 mg
446410 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 446450 Supplier R&D Systems™ Supplier No. 4464/50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. Histone demethylase inhibitor; cell permeable

IOX 1 is a histone demethylase JMJD inhibitor (IC50 values are 0.12, 0.17, 0.2, 0.3, 0.6 and 1 μM for JMJD3, JMJD1A, JMJD2A, JMJD2E, JMJD2C and UTX respectively). Cell permeable; it inhibits JMJD2A-mediated H3K9me3 demethylation in HeLa cells. IOX 1 suppresses IL-17 expression in mouse and human CD4+ T cells by targeting TET2. It also reduces infiltration of Th17 cells in an in vivo model of intraocular inflammation.

Chemical Identifiers

CAS 5852-78-8
Molecular Formula C10H7NO3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 189.17
Purity >99%
Molecular Weight (g/mol) 189.17
Chemical Name or Material IOX 1
Recommended Storage Store at +4°C