accessibility menu, dialog, popup

UserName

NOHOOH

CAS RN 5852-78-8

IUPAC Name: 8-hydroxyquinoline-5-carboxylic acid
Synonyms: 8-hydroxyquinoline-5-carboxylic acid
Molecular Weight (g/mol): 189.17
Molecular Formula: C10H7NO3
InChi Key: JGRPKOGHYBAVMW-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(O)C2=NC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results