missing translation for 'onlineSavingsMsg'
Learn More

Ion exchanger Dowex™ 1-X8, Cl- form, EMD-Millipore

Catalog No. M1052420250
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1052420250 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1052420250 Supplier Technidata Supplier No. 1.05242.0250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Strongly basic anion exchangers

Chemical Identifiers

CAS 60177-39-1
Molecular Formula C22H31ClN
Molecular Weight (g/mol) 344.95

Specifications

Density Density: 1.040g/cm3 (20°)
Quantity 250 mL
Chemical Name or Material Ion exchanger Dowex™ 1-X8
Recommended Storage Store at 5° to 30°C