missing translation for 'onlineSavingsMsg'
Learn More

Iodotrimethylsilane, 97%, stab. with copper

Catalog No. AAA1290230
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1290230 250 g
AAA1290206 5 g
AAA1290214 25 g
AAA1290222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1290230 Supplier Thermo Scientific Chemicals Supplier No. A1290230
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 16029-98-4 | C3H9ISi | 200.094 g/mol

Iodotrimethylsilane is used for the introduction of trimethylsilyl group in organic synthesis. It is also useful for gas chromatography analysis by converting alcohol into a silyl ether derivative, thereby making it more volatile than the original molecule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodotrimethylsilane is used for the introduction of trimethylsilyl group in organic synthesis. It is also useful for gas chromatography analysis by converting alcohol into a silyl ether derivative, thereby making it more volatile than the original molecule.

Solubility
Miscible with water.

Notes
Moisture and light Sensitive. Incompatible withstrong acids, strong bases, strong oxidizing agents and reacts violently with water.

Chemical Identifiers

CAS 16029-98-4
Molecular Formula C3H9ISi
Molecular Weight (g/mol) 200.094
MDL Number MFCD00001028
InChI Key CSRZQMIRAZTJOY-UHFFFAOYSA-N
Synonym trimethylsilyl iodide, trimethyliodosilane, silane, iodotrimethyl, iodo trimethyl silane, trimethylsilyliodide, tms-iodide, iodotrimethysilane, iodotrimetylsilane, tms iodide, iodo trimethylsilane
PubChem CID 85247
IUPAC Name iodo(trimethyl)silane
SMILES C[Si](C)(C)I

Specifications

Boiling Point 105°C to 106°C
Quantity 250 g
Linear Formula (CH3)3SiI
UN Number UN2924
Beilstein 1731136
Flash Point −2°C (28°F)
Solubility Information Miscible with water.
Formula Weight 200.09
Percent Purity 97%
Refractive Index 1.476
Sensitivity Light and moisture sensitive
Density 1.44
Chemical Name or Material Iodotrimethylsilane, stabilized with copper
Show More Show Less
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 240-171-0
TSCA Yes
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only