missing translation for 'onlineSavingsMsg'
Learn More

Iodopyrazine, 97%

Catalog No. AC397530050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397530050 5 g Glass bottle
AC397530010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397530050 Supplier Thermo Scientific Chemicals Supplier No. 397530050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 32111-21-0 | C4H3IN2 | 205.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32111-21-0
Molecular Formula C4H3IN2
Molecular Weight (g/mol) 205.98
MDL Number MFCD01319019
InChI Key OYWPFIUVDKHHGQ-UHFFFAOYSA-N
Synonym iodopyrazine, 2-iodo-pyrazine, 2-iodo-1,3-diazine, iodo-pyrazine, 2-iodo pyrazine, pyrazine, 2-iodo, acmc-1ad2s
PubChem CID 642841
IUPAC Name 2-iodopyrazine
SMILES C1=CN=C(C=N1)I

Specifications

Color White to Yellow
Density 2.0860g/mL
Boiling Point 109°C to 110°C (34.0 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6550 to 1.6580 (20°C, 589nm)
Quantity 5 g
Specific Gravity 2.086
Solubility Information Solubility in water: .. Other solubilities: soluble in ether,acetone,benzene,hexanes
Formula Weight 205.98
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Iodopyrazine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only