missing translation for 'onlineSavingsMsg'
Learn More

Iodomethane Solution, Certified Reference Material, 2,000 μg/mL in Methanol: Water (4:1), MilliporeSigma™ Supelco™

Catalog No. 111020502
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020502 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020502 Supplier Merck Emd Millipore Supplier No. 506052

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

Packaging Ampule
CAS 74-88-4
Product Line TraceCERT
Concentration 2,000 μg/mL in methanol: water (4:1)
Linear Formula CH3I
MDL Number MFCD00001073
Synonym Methyl iodide
Type Food and Cosmetic Component Standard
Quantity 1 mL
Formula Weight 141.94
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC