missing translation for 'onlineSavingsMsg'
Learn More

Iodoethane (stabilized with Copper chip) 99.0+%, TCI America™

Catalog No. I0058100G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0058100G 100 g
I005825G 25 g
I0058500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0058100G Supplier TCI America Supplier No. I0058100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-03-6
Molecular Formula C2H5I
Molecular Weight (g/mol) 155.966
MDL Number MFCD00001091
InChI Key HVTICUPFWKNHNG-UHFFFAOYSA-N
Synonym ethyl iodide, ethane, iodo, monoiodoethane, hydriodic ether, ethyliodide, jodethan czech, ethyljodid, jodethan, iodo-ethane, ethyljodid czech
PubChem CID 6340
ChEBI CHEBI:42487
IUPAC Name iodoethane
SMILES CCI

Specifications

Melting Point -108°C
Color Colorless
Boiling Point 72°C
Quantity 100 g
UN Number 1993
Formula Weight 155.97
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Iodoethane (stabilized with Copper chip)
EINECSNumber (2)-0079
RTECSNumber KI4750000
TSCA Yes