missing translation for 'onlineSavingsMsg'
Learn More

Iodine (Iodine-Iodide), 0.100 N (N/10), Ricca Chemical

Catalog No. 400032
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
400032 1 L Amber Glass Bottle
400016 500 mL Amber Glass Bottle
40001 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 400032 Supplier Ricca Chemical Company Supplier No. 400032
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7553-56-2 , 7681-11-0 , 7732-18-5
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name diiodine
SMILES II

Specifications

Color Brown
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.100N
Packaging Amber Glass Bottle
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Iodine (Iodine-Iodide) Solution, Iodine-Iodide