missing translation for 'onlineSavingsMsg'
Learn More

Iodine (Iodine-Iodide), 0.100 N (N/10), Ricca Chemical

Supplier:  Ricca Chemical Company RI40001

 View more versions of this product

Catalog No. 4000-1


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Iodine (Iodine-Iodide) Solution
Iodine-Iodide
I2
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
II
253.81
CHEBI:17606
0.100N
4 L
Brown
7553-56-2 , 7681-11-0 , 7732-18-5
MFCD00011355 MFCD00164163
PNDPGZBMCMUPRI-UHFFFAOYSA-N
diiodine
807
Laboratory
Amber Glass Bottle
Liquid

Chemical Identifiers

7553-56-2
253.81
PNDPGZBMCMUPRI-UHFFFAOYSA-N
807
diiodine
I2
MFCD00011355 MFCD00164163
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
CHEBI:17606
II
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions