missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine (Iodine-Iodide), 0.0250 N (N/40), 1 mL = 0.4008 mg S2-, Ricca Chemical
Specifications
Specifications
| Color | Brown |
| CAS Min % | 0.25, 96.0, 1.75 |
| CAS Max % | 0.26, 100.0, 1.82 |
| Equivalence | 1 mL = 0.4008 mg S |
| Grade | Laboratory |
| Packaging | Amber Glass Bottle |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Concentration or Composition Notes | 0.0250Normal |
| Chemical Name or Material | Iodine, Iodine-Iodide |