missing translation for 'onlineSavingsMsg'
Learn More

Iodine, Certified, 0.0250N ±0.0003N (0.0125M), LabChem™

Catalog No. p-5388299
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC156201 500 mL Amber Glass
LC156202 1 L Amber Glass
LC156204 4 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC156201 Supplier LabChem Supplier No. LC156201
Only null left

Chemical Identifiers

CAS 7553-56-2 , 7732-18-5 , 7681-11-0 , 7647-01-0
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name diiodine
SMILES II

Specifications

Color Red through Brown
CAS Max % 0.32, 99.02, 0.64, 0.02
Physical Form Liquid
Quantity 500 mL
Linear Formula I2
Solubility Information Soluble in water
Formula Weight 253.809
Grade Certified
Concentration or Composition (by Analyte or Components) 0.0250N ±0.0003N (0.0125M)
Identification Passes Test
Packaging Amber Glass
Chemical Name or Material Iodine
Show More Show Less
Hazard Statement GHS H Statement
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature