missing translation for 'onlineSavingsMsg'
Learn More

Iodine, Certified, 0.0200N ±0.0002N (0.01M), LabChem™

Catalog No. LC156102
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC156102 1 L Amber Glass
LC156104 4 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC156102 Supplier LabChem Supplier No. LC156102
Only null left

Chemical Identifiers

CAS 7553-56-2 , 7732-18-5 , 7681-11-0 , 7647-01-0
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name diiodine
SMILES II

Specifications

Color Red through Brown
CAS Max % 0.26, 99.22, 0.5, 0.02
Physical Form Liquid
Quantity 1 L
Linear Formula I2
Solubility Information Soluble in water
Formula Weight 253.809
Grade Certified
Concentration or Composition (by Analyte or Components) 0.0200N ±0.0002N (0.01M)
Identification Passes Test
Packaging Amber Glass
Chemical Name or Material Iodine
Show More Show Less
Hazard Statement GHS H Statement
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature