missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine, Certified, 0.0167N ±0.0001N (0.0084M), LabChem™
Specifications
Specifications
| Color | Red through Brown |
| CAS Max % | 0.21, 99.35, 0.42, 0.02 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Linear Formula | I2 |
| Solubility Information | Soluble in water |
| Formula Weight | 253.809 |
| Grade | Certified |
| Concentration or Composition (by Analyte or Components) | 0.0167N ±0.0001N (0.0084M) |
| Identification | Passes Test |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Harmful to aquatic life. |
| Precautionary Statement | GHS P Statement Avoid release to the environment. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |