missing translation for 'onlineSavingsMsg'
Learn More

Iodine, Certified, 0.0100N ±0.0001N (0.005M), LabChem™

Catalog No. LC156002
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass
Pvc Coated
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC156002 1 L Pvc Coated
LC156001 500 mL Amber Glass
LC156004 4 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC156002 Supplier LabChem Supplier No. LC156002

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7553-56-2 | I2 | 253.81 g/mol

Chemical Identifiers

CAS 7553-56-2 , 7732-18-5 , 7681-11-0 , 7647-01-0
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
IUPAC Name diiodine
SMILES II

Specifications

CAS Max % 99.6, 0.25, 0.13, 0.02
Physical Form Liquid
Quantity 1 L
Linear Formula I2
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Solubility Information Soluble in water
Formula Weight 253.809
Grade Certified
Packaging Pvc Coated
Chemical Name or Material Iodine, 0.01N (0.005M)
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.