missing translation for 'onlineSavingsMsg'
Learn More

Iodine, Certified, 0.0100N ±0.0001N (0.005M), LabChem™

Catalog No. LC156001
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass
Pvc Coated
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC156001 500 mL Amber Glass
LC156002 1 L Pvc Coated
LC156004 4 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC156001 Supplier LabChem Supplier No. LC156001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7553-56-2 | I2 | 253.81 g/mol

Chemical Identifiers

CAS 7553-56-2 , 7732-18-5 , 7681-11-0 , 7647-01-0
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name diiodine
SMILES II

Specifications

Color Red through Brown
CAS Max % 0.13, 99.6, 0.25, 0.02
Physical Form Liquid
Quantity 500 mL
Linear Formula I2
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
Solubility Information Soluble in water
Formula Weight 253.809
Grade Certified
Concentration or Composition (by Analyte or Components) 0.0100N ±0.0001N (0.005M)
Identification Passes Test
Packaging Amber Glass
Chemical Name or Material Iodine
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature